Assay Details
Target Gene Details
Entrez Gene ID: | 51938 |
Gene Name: | coiled-coil domain containing 39 |
Gene Aliases: |
4921507O14Rik, AI447426, AI844750, D3Ertd789e, b2b1304Clo, b2b1735Clo, b2b2025.1Clo |
Location: |
Chr.3:33810757-33844322 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc39 | NM_026222.2 | 4 | 561 | NP_080498.1 |
| AK014839.1 | 4 | 553 | BAB29574.1 | |
| AK029948.1 | 4 | 517 | ||
| AK030087.1 | 4 | 528 | BAC26777.1 | |
| AK083226.1 | 4 | 433 | ||
| BC138917.1 | 4 | 523 | ||
| BC138920.1 | 4 | 523 |

Genomic Map