Assay Details
Target Gene Details
Entrez Gene ID: | 18033 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 |
Gene Aliases: |
NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105 |
Location: |
Chr.3:135584655-135691547 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkb1 | NM_008689.2 | 9 | 1144 | NP_032715.2 |
| AB119195.1 | 7 | 642 | BAC84979.1 | |
| AB221614.1 | 7 | 649 | ||
| AK052726.1 | 4 | 452 | BAC35117.1 | |
| AY521462.1 | 7 | 642 | AAS00547.1 | |
| AY521463.1 | 7 | 642 | AAS00548.1 | |
| BC050841.1 | 9 | 1144 | AAH50841.1 | |
| BC138535.1 | 8 | 721 | ||
| BC138536.1 | 8 | 721 | ||
| M57999.1 | 9 | 933 | AAA40415.1 |
More Information
Set Membership: |
|

Genomic Map