Assay Details
Target Gene Details
Entrez Gene ID: | 74446 |
Gene Name: | solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1 |
Gene Aliases: |
1700094G20Rik, 4933424B12Rik, 4933425K02Rik, Nhedc1, mtsNHE |
Location: |
Chr.3:135347994-135397827 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9b1 | NM_028946.3 | 6 | 1133 | NP_083222.2 |
| AK016883.1 | 6 | 985 | BAC25498.1 | |
| AK016917.1 | 6 | 1113 | BAB30495.1 | |
| AK029525.1 | 6 | 1134 | BAC26494.1 | |
| BC025002.1 | 5 | 1055 | AAH25002.1 | |
| EU846100.1 | 5 | 664 |

Genomic Map