Assay Details
Target Gene Details
Entrez Gene ID: | 381489 |
Gene Name: | relaxin/insulin-like family peptide receptor 1 |
Gene Aliases: |
Gm1018, Lgr7 |
Location: |
Chr.3:79641600-79737951 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rxfp1 | NM_212452.1 | NP_997617.1 | ||
| AK137655.1 | 1 | 74 | ||
| AY509975.1 | AAR97515.1 |
More Information
Set Membership: |
|

Genomic Map