Assay Details
Target Gene Details
Entrez Gene ID: | 28146 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
D3Ucla1, Ramp4 |
Location: |
Chr.3:58521969-58525884 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
More Information
Set Membership: |
|

Genomic Map