Assay Details
Target Gene Details
Entrez Gene ID: | 97112 |
Gene Name: | NMD3 homolog (S. cerevisiae) |
Gene Aliases: |
C87860 |
Location: |
Chr.3:69722055-69749047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmd3 | NM_133787.2 | NP_598548.1 | ||
| AK088667.1 | BAC40491.1 | |||
| AK145248.1 | BAE26326.1 | |||
| AK166283.1 | BAE38680.1 | |||
| BC003842.1 | AAH03842.1 |

Genomic Map