Assay Details
Target Gene Details
Entrez Gene ID: | 229700 |
Gene Name: | RNA binding motif protein 15 |
Gene Aliases: |
C230088J01Rik, mKIAA1438 |
Location: |
Chr.3:107326110-107333289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm15 | NM_001045807.1 | NP_001039272.1 | ||
| AK077048.1 | ||||
| AK168242.1 | BAE40192.1 | |||
| AK168936.1 | ||||
| BC028452.1 | ||||
| BC051409.1 | AAH51409.1 | |||
| BC057038.1 | AAH57038.1 | |||
| BC080828.1 | AAH80828.1 | |||
| BC120590.1 | ||||
| BC137741.1 |
More Information
Set Membership: |
|

Genomic Map