Assay Details
Target Gene Details
Entrez Gene ID: | 81004 |
Gene Name: | transducin (beta)-like 1X-linked receptor 1 |
Gene Aliases: |
8030499H02Rik, A630076E03Rik, AW539987, C21, C230089I12Rik, DC42, Ira1, TBLR1 |
Location: |
Chr.3:22076593-22216594 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbl1xr1 | NM_030732.3 | NP_109657.2 | ||
| AF268195.1 | AAG44738.1 | |||
| AK029595.1 | BAC26526.1 | |||
| AK033347.1 | BAC28241.1 | |||
| AK036064.1 | BAC29294.1 | |||
| AK136255.1 | BAE22898.1 |

Genomic Map