Assay Details
Target Gene Details
Entrez Gene ID: | 20201 |
Gene Name: | S100 calcium binding protein A8 (calgranulin A) |
Gene Aliases: |
60B8Ag, AI323541, B8Ag, CFAg, CP-10, Caga, MRP8, p8 |
Location: |
Chr.3:90669071-90670034 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a8 | NM_013650.2 | NP_038678.1 | ||
| AK157837.1 | BAE34219.1 | |||
| AV007095.1 | ||||
| BC078629.1 | AAH78629.1 | |||
| M83218.1 | AAB07229.1 | |||
| S57123.1 | AAB25840.1 |
More Information
Set Membership: |
|

Genomic Map