Assay Details
Target Gene Details
Entrez Gene ID: | 17210 |
Gene Name: | myeloid cell leukemia sequence 1 |
Gene Aliases: |
AW556805, Mcl-1 |
Location: |
Chr.3:95658368-95663179 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcl1 | NM_008562.3 | 2 | 801 | NP_032588.1 |
| AK010424.1 | BAB26927.1 | |||
| AK159504.1 | 2 | 713 | BAE35137.1 | |
| AK160594.1 | BAE35901.1 | |||
| AK215970.1 | ||||
| BC003839.1 | 2 | 730 | AAH03839.1 | |
| BC005427.1 | 2 | 723 | AAH05427.1 | |
| BC021638.1 | 2 | 740 | AAH21638.1 | |
| GU182318.1 | 3 | 527 | ||
| U35623.1 | 2 | 713 | AAC31790.1 |

Genomic Map