Assay Details
Target Gene Details
Entrez Gene ID: | 26570 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 11 |
Gene Aliases: |
9930009M05Rik, AI451155, sut, xCT |
Location: |
Chr.3:50364936-50499087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a11 | NM_011990.2 | NP_036120.1 | ||
| AB022345.1 | BAA77220.1 | |||
| AK037490.1 | ||||
| AK037742.1 | BAC29863.1 | |||
| AK076322.1 | ||||
| AK089202.1 | BAC40792.1 | |||
| AK162233.1 | ||||
| AK163168.1 | BAE37218.1 | |||
| AK165599.1 | BAE38284.1 | |||
| AK171192.1 | BAE42302.1 | |||
| AK171252.1 | BAE42344.1 | |||
| AY766236.1 | AAX10866.1 | |||
| BC141402.1 | ||||
| BC141408.1 |
More Information
Set Membership: |
|

Genomic Map