Assay Details
Target Gene Details
| Entrez Gene ID: | 100505386 | 
| Gene Name: | predicted gene, 21949 | 
| Gene Aliases: | Iqcj-schip1, Schip-1-iqcj, Schip1 | 
| Location: | Chr.3:67892220-68626482 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Exon 3 - Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Gm21949 | NM_001113419.2 | NP_001106890.1 | ||
| KJ941154.1 | 
Target Gene Details
| Entrez Gene ID: | 208426 | 
| Gene Name: | IQ motif containing J | 
| Gene Aliases: | D230050A05, EG208426 | 
| Location: | Chr.3:67892220-68056593 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Exon 3 - Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Iqcj | NM_177585.3 | NP_808253.1 | ||
| AK052136.1 | BAC34854.1 | |||
| BC107305.1 | AAI07306.1 | |||
| BC107306.1 | AAI07307.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic