Assay Details
Target Gene Details
Entrez Gene ID: | 70099 |
Gene Name: | structural maintenance of chromosomes 4 |
Gene Aliases: |
2500002A22Rik, C79747, SMC-4, Smc4l1 |
Location: |
Chr.3:69004972-69034623 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc4 | NM_133786.3 | 21 | 3360 | NP_598547.1 |
| AJ534940.2 | 20 | 3236 | CAD59183.1 | |
| BC061481.1 | 21 | 3335 | AAH61481.1 | |
| BC062939.1 | 21 | 3360 | AAH62939.1 |

Genomic Map