Assay Details
Target Gene Details
Entrez Gene ID: | 59006 |
Gene Name: | myozenin 2 |
Gene Aliases: |
1110012I24Rik, Fatz-2, Myozl2 |
Location: |
Chr.3:123006206-123035001 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myoz2 | NM_021503.2 | 1 | 128 | NP_067478.1 |
| AJ252148.1 | 1 | 90 | CAB92972.1 | |
| AK003634.1 | 1 | 80 | ||
| AK085927.1 | 1 | 93 | ||
| AY013296.1 | 1 | 104 | AAG38939.1 | |
| BC024360.1 | 1 | 90 | AAH24360.1 |
More Information
Set Membership: |
|

Genomic Map