Assay Details
Target Gene Details
Entrez Gene ID: | 18167 |
Gene Name: | neuropeptide Y receptor Y2 |
Gene Aliases: |
NPY2-R |
Location: |
Chr.3:82538383-82548989 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npy2r | NM_001205099.1 | 1 | 210 | NP_001192028.1 |
| NM_008731.3 | 1 | 210 | NP_032757.2 | |
| AK018543.1 | 1 | 187 | ||
| AK049892.1 | 1 | 198 | BAC33975.1 | |
| BC104490.1 | 1 | 85 | AAI04491.1 | |
| BC104733.1 | 1 | 85 | AAI04734.1 | |
| BC104734.1 | 1 | 85 | AAI04735.1 | |
| D86238.1 | 1 | 216 | BAA13050.1 |
More Information
Set Membership: |
|

Genomic Map