Assay Details
Target Gene Details
Entrez Gene ID: | 229214 |
Gene Name: | pyroglutamylated RFamide peptide receptor |
Gene Aliases: |
AQ27, Gpr103, SP9155 |
Location: |
Chr.3:36179426-36222275 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qrfpr | NM_198192.2 | NP_937835.1 | ||
| AB109631.1 | BAC98940.1 | |||
| AK034431.1 | BAE20492.1 | |||
| AK043649.1 | BAE20642.1 | |||
| AK044246.1 | ||||
| AK142880.1 | BAE25217.1 | |||
| BC096610.1 | AAH96610.1 |
More Information
Set Membership: |
|

Genomic Map