Assay Details
Target Gene Details
Entrez Gene ID: | 66868 |
Gene Name: | major facilitator superfamily domain containing 1 |
Gene Aliases: |
1200003O06Rik |
Location: |
Chr.3:67582768-67604231 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd1 | NM_025813.3 | 1 | 61 | NP_080089.1 |
| AK004586.1 | 1 | 62 | BAB23391.1 | |
| AK032688.1 | ||||
| AK075859.1 | 1 | 91 | BAC36010.1 | |
| AK135235.1 | 1 | 62 | BAE22467.1 | |
| AK136108.1 | BAE22824.1 | |||
| AK152444.1 | 1 | 83 | BAE31224.1 | |
| AK159554.1 | 1 | 89 | BAE35179.1 | |
| AK166645.1 | ||||
| BC024891.1 | AAH24891.1 |

Genomic Map