Assay Details
Target Gene Details
Entrez Gene ID: | 19892 |
Gene Name: | retinal pigment epithelium 65 |
Gene Aliases: |
65kDa, A930029L06Rik, LCA2, Mord1, RP20, rd12 |
Location: |
Chr.3:159599075-159625307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpe65 | NM_029987.2 | NP_084263.2 | ||
| AF410461.1 | AAL01119.1 | |||
| AK020906.1 | BAB32248.1 | |||
| BC130028.1 | ||||
| CJ133618.1 |
More Information
Set Membership: |
|

Genomic Map