Assay Details
Target Gene Details
Entrez Gene ID: | 20333 |
Gene Name: | SEC22 vesicle trafficking protein homolog B (S. cerevisiae) |
Gene Aliases: |
4930564D15Rik, AA517334, AI480645, C81333, ERS-24, Sec22l1 |
Location: |
Chr.3:97901227-97922318 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec22b | NM_011342.4 | NP_035472.1 | ||
AK010363.1 | ||||
AK017237.1 | BAB30646.1 | |||
AK034973.1 | BAC28899.1 | |||
AK037918.1 | BAC29901.1 | |||
AK088514.1 | BAC40397.1 | |||
AK089928.1 | BAC40999.1 | |||
AK134215.1 | BAE22055.1 | |||
AK178690.1 | ||||
AK190268.1 | ||||
BC009024.1 | AAH09024.1 | |||
U91538.1 | AAC53130.1 |
