Assay Details
Target Gene Details
Entrez Gene ID: | 66377 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1 |
Gene Aliases: |
2310016K22Rik, KFYI |
Location: |
Chr.3:51405479-51408955 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufc1 | NM_025523.1 | NP_079799.1 | ||
| AK003365.1 | BAB22739.1 | |||
| AK009393.1 | BAB26262.1 | |||
| AK018705.1 | BAB31356.1 | |||
| BC092141.1 | AAH92141.1 |
More Information
Set Membership: |
|

Genomic Map