Assay Details
Target Gene Details
Entrez Gene ID: | 74776 |
Gene Name: | pyrophosphatase (inorganic) 2 |
Gene Aliases: |
1110013G13Rik, Sid6306 |
Location: |
Chr.3:133309159-133378235 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppa2 | NM_001293641.1 | NP_001280570.1 | ||
| NM_146141.2 | NP_666253.1 | |||
| AK003660.1 | BAB22922.1 | |||
| AK090384.1 | BAC41194.1 | |||
| AK143475.1 | BAE25392.1 | |||
| BC011417.1 | AAH11417.1 |
More Information
Set Membership: |
|

Genomic Map