Assay Details
Target Gene Details
Entrez Gene ID: | 72175 |
Gene Name: | major facilitator superfamily domain containing 8 |
Gene Aliases: |
2810423E13Rik, AI836898, AV142426, Cln7 |
Location: |
Chr.3:40818112-40846856 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd8 | NM_028140.4 | NP_082416.2 | ||
| AK013144.1 | BAB28676.1 | |||
| AK032080.1 | BAC27687.1 | |||
| AK035291.1 | ||||
| AK077957.1 | BAC37083.1 | |||
| BC010483.1 | AAH10483.1 | |||
| BC113183.1 | ||||
| BC113790.1 |

Genomic Map