Assay Details
Target Gene Details
Entrez Gene ID: | 18770 |
Gene Name: | pyruvate kinase liver and red blood cell |
Gene Aliases: |
Pk-1, Pk1, R-PK |
Location: |
Chr.3:89136122-89146600 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pklr | NM_001099779.1 | NP_001093249.1 | ||
| NM_013631.2 | NP_038659.2 | |||
| AK149508.1 | BAE28927.1 | |||
| AK149524.1 | BAE28937.1 | |||
| BC152327.1 | ||||
| BY107231.1 | ||||
| D63764.1 | BAA23642.1 | |||
| S79731.1 | AAB35435.1 |
More Information
Set Membership: |
|

Genomic Map