Assay Details
Target Gene Details
Entrez Gene ID: | 20132 |
Gene Name: | retinal pigment epithelium derived rhodopsin homolog |
Gene Aliases: |
- |
Location: |
Chr.3:129808575-129822505 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrh | NM_009102.3 | NP_033128.1 | ||
| AF012271.1 | AAC53344.1 | |||
| AK021379.1 | BAB32391.1 | |||
| BC046288.1 | AAH46288.1 |
More Information
Set Membership: |
|

Genomic Map