Assay Details
Target Gene Details
Entrez Gene ID: | 18033 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 |
Gene Aliases: |
NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105 |
Location: |
Chr.3:135584655-135691547 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkb1 | NM_008689.2 | 15 | 1932 | NP_032715.2 |
| AB119195.1 | 13 | 1430 | BAC84979.1 | |
| AB221614.1 | 13 | 1437 | ||
| AK089660.1 | 3 | 306 | BAE43399.1 | |
| AK146389.1 | 1 | 152 | ||
| AK159423.1 | 3 | 363 | ||
| AY521462.1 | 13 | 1430 | AAS00547.1 | |
| AY521463.1 | 13 | 1430 | AAS00548.1 | |
| BC138535.1 | 14 | 1509 | ||
| BC138536.1 | 14 | 1509 | ||
| L28117.1 | 3 | 349 | ||
| M57999.1 | 15 | 1721 | AAA40415.1 | |
| S66656.1 | 3 | 476 | AAB28573.1 | |
| S89033.1 | 3 | 524 | AAB21851.1 |
More Information
Set Membership: |
|

Genomic Map