Assay Details
Target Gene Details
Entrez Gene ID: | 18033 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 |
Gene Aliases: |
NF-KB1, NF-kappaB, NF-kappaB1, p105, p50, p50/p105 |
Location: |
Chr.3:135584655-135691547 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkb1 | NM_008689.2 | 23 | 2953 | NP_032715.2 |
| AB119195.1 | 21 | 2451 | BAC84979.1 | |
| AB221614.1 | 21 | 2458 | ||
| AK089660.1 | 11 | 1327 | BAE43399.1 | |
| AK146389.1 | 9 | 1172 | ||
| AK157915.1 | 8 | 953 | BAE34261.1 | |
| AK159423.1 | 11 | 1384 | ||
| AY521462.1 | 21 | 2451 | AAS00547.1 | |
| AY521463.1 | 21 | 2451 | AAS00548.1 | |
| BC138535.1 | 22 | 2530 | ||
| BC138536.1 | 22 | 2530 | ||
| L28117.1 | 11 | 1351 | ||
| L28118.1 | 7 | 838 | ||
| M57999.1 | 23 | 2742 | AAA40415.1 | |
| S66656.1 | 11 | 1497 | AAB28573.1 | |
| S89033.1 | 11 | 1545 | AAB21851.1 |
More Information
Set Membership: |
|

Genomic Map