Assay Details
Target Gene Details
Entrez Gene ID: | 66880 |
Gene Name: | arginine/serine-rich coiled-coil 1 |
Gene Aliases: |
1200013F24Rik, SRrp53 |
Location: |
Chr.3:66985579-67358403 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rsrc1 | NM_025822.3 | NP_080098.1 | ||
AK004742.2 | BAB23523.1 | |||
AK005242.1 | ||||
AK045433.1 | BAC32364.1 | |||
AK168539.1 | BAE40417.1 | |||
AK169340.1 | BAE41092.1 | |||
BC038303.1 | AAH38303.1 | |||
BC043045.1 | AAH43045.1 | |||
BC083123.1 | AAH83123.1 |
