Assay Details
Target Gene Details
Entrez Gene ID: | 12353 |
Gene Name: | carbonic anhydrase 6 |
Gene Aliases: |
Ca6, DOC1 |
Location: |
Chr.4:150187015-150201135 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car6 | NM_009802.2 | 2 | 150 | NP_033932.2 |
| AF079834.1 | 1 | 240 | AAD12539.1 | |
| AF079835.1 | 2 | 177 | AAD12540.1 | |
| AK132599.1 | 2 | 151 | BAE21254.1 | |
| AK161216.1 | 2 | 146 | BAE36243.1 | |
| BC006726.1 | 2 | 237 | ||
| BC049973.1 | 2 | 148 | AAH49973.1 | |
| BC055437.1 | 2 | 136 | AAH55437.1 |

Genomic Map