Assay Details
Target Gene Details
Entrez Gene ID: | 23833 |
Gene Name: | CD52 antigen |
Gene Aliases: |
AI463198, B7, B7-Ag, CAMPATH-1, CLS1, MB7 |
Location: |
Chr.4:134093538-134095073 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd52 | NM_013706.2 | NP_038734.1 | ||
| AK088161.1 | BAC40183.1 | |||
| AK155728.1 | BAE33405.1 | |||
| BC027495.1 | AAH27495.1 | |||
| M55561.1 | AAA39889.1 | |||
| X76696.1 | CAA54125.1 |

Genomic Map