Assay Details
Target Gene Details
Entrez Gene ID: | 66291 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
1810030N24Rik, 2810406B13Rik |
Location: |
Chr.4:34768664-34778396 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smim8 | NM_025471.2 | 3 | 377 | NP_079747.1 |
| AK005941.1 | 3 | 380 | BAB24328.1 | |
| AK007303.1 | 2 | 135 | BAB24944.1 | |
| AK007660.1 | 2 | 155 | BAB25170.1 | |
| AK021236.1 | 2 | 172 | BAB32341.1 | |
| AK079613.1 | 2 | 166 | ||
| BC027508.1 | 2 | 125 | AAH27508.1 |

Genomic Map