Assay Details
Target Gene Details
Entrez Gene ID: | 230393 |
Gene Name: | focadhesin |
Gene Aliases: |
Kiaa1797, mKIAA1797 |
Location: |
Chr.4:88094630-88411011 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Focad | NM_001081184.1 | 21 | 2632 | NP_001074653.1 |
| BC057079.1 | 10 | 1110 | AAH57079.1 |

Genomic Map