Assay Details
Target Gene Details
Entrez Gene ID: | 230099 |
Gene Name: | carbonic anhydrase 9 |
Gene Aliases: |
CAIX, Ca9, MN/CA9 |
Location: |
Chr.4:43507026-43513810 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car9 | NM_139305.2 | NP_647466.2 | ||
| AB086322.1 | BAC00816.1 | |||
| AJ245857.1 | CAC80975.1 | |||
| AK136579.1 | BAE23060.1 | |||
| AK137719.1 | BAE23474.1 | |||
| AK166403.1 | BAE38755.1 | |||
| BC120544.1 | ||||
| BC120546.1 | ||||
| EF122497.1 |

Genomic Map