Assay Details
Target Gene Details
Entrez Gene ID: | 26885 |
Gene Name: | caspase 8 associated protein 2 |
Gene Aliases: |
AA387232, D4Ertd659e, FLASH, mKIAA1315 |
Location: |
Chr.4:32615473-32653271 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp8ap2 | NM_001122978.1 | NP_001116450.1 | ||
| NM_011997.2 | NP_036127.2 | |||
| AF132726.1 | AAD29045.1 | |||
| AK011818.1 | BAB27860.1 | |||
| BC138022.1 | ||||
| BC138041.1 |
More Information
Set Membership: |
|

Genomic Map