Assay Details
Target Gene Details
Entrez Gene ID: | 11876 |
Gene Name: | artemin |
Gene Aliases: |
neublastin |
Location: |
Chr.4:117926160-117930791 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Artn | NM_001284191.1 | NP_001271120.1 | ||
| NM_001284192.1 | NP_001271121.1 | |||
| NM_001284193.1 | NP_001271122.1 | |||
| NM_009711.4 | NP_033841.1 | |||
| AF109402.1 | AAC98691.1 | |||
| AK015393.1 | BAB29827.1 | |||
| AK053914.1 | BAC35590.1 | |||
| AK145190.1 | ||||
| AK156507.1 | BAE33737.1 | |||
| AK166013.1 | ||||
| BC104328.1 | 1 | 615 | AAI04329.1 | |
| BC104329.1 | AAI04330.1 |
More Information
Set Membership: |
|

Genomic Map