Assay Details
Target Gene Details
Entrez Gene ID: | 12537 |
Gene Name: | cyclin-dependent kinase 11B |
Gene Aliases: |
AA989746, CDK11-p110, CDK11-p46, CDK11-p58, Cdc11b, Cdc2l1, Cdc2l2, Cdk11, p58 |
Location: |
Chr.4:155624846-155649935 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk11b | NM_007661.3 | 13 | 1500 | NP_031687.2 |
| AK077668.1 | 13 | 1501 | BAC36942.1 | |
| AK147133.1 | 13 | 1478 | BAE27703.1 | |
| BC025058.1 | 2 | 150 | AAH25058.1 | |
| BC052920.1 | 13 | 1578 | AAH52920.1 | |
| L37092.1 | 12 | 2072 | AAA66169.1 | |
| M58633.1 | 8 | 843 | AAA03518.1 |
More Information
Set Membership: |
|

Genomic Map