Assay Details
Target Gene Details
Entrez Gene ID: | 22253 |
Gene Name: | unc-5 homolog C (C. elegans) |
Gene Aliases: |
B130051O18Rik, Unc5h3, rcm |
Location: |
Chr.3:141465564-141834924 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc5c | NM_001293561.1 | NP_001280490.1 | ||
| NM_009472.4 | NP_033498.1 | |||
| AK031655.1 | BAC27495.1 | |||
| AK141181.1 | BAE24575.1 | |||
| AK142411.1 | BAE25058.1 | |||
| AK142974.1 | BAE25236.1 | |||
| AK164239.1 | BAE37698.1 | |||
| BC115772.1 | ||||
| U72634.1 | AAB54103.1 |
More Information
Set Membership: |
|

Genomic Map