Assay Details
Target Gene Details
Entrez Gene ID: | 12537 |
Gene Name: | cyclin-dependent kinase 11B |
Gene Aliases: |
AA989746, CDK11-p110, CDK11-p46, CDK11-p58, Cdc11b, Cdc2l1, Cdc2l2, Cdk11, p58 |
Location: |
Chr.4:155624846-155649935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk11b | NM_007661.3 | NP_031687.2 | ||
| AK008283.1 | BAB25574.1 | |||
| AK077668.1 | BAC36942.1 | |||
| AK135402.1 | BAE22520.1 | |||
| AK147133.1 | BAE27703.1 | |||
| AK171785.1 | 1 | 681 | ||
| BC052920.1 | AAH52920.1 | |||
| L37092.1 | 1 | 697 | AAA66169.1 |
More Information
Set Membership: |
|

Genomic Map