Assay Details
Target Gene Details
Entrez Gene ID: | 329954 |
Gene Name: | cation channel, sperm associated 4 |
Gene Aliases: |
4933412L05 |
Location: |
Chr.4:134211968-134227434 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsper4 | NM_001130030.1 | NP_001123502.1 | ||
| NM_177866.4 | NP_808534.1 | |||
| AK077145.1 | BAC36641.1 | |||
| BC108974.2 | AAI08975.1 | |||
| BC108975.1 | AAI08976.1 | |||
| EU781510.1 | ||||
| EU781511.1 |

Genomic Map