Assay Details
Target Gene Details
Entrez Gene ID: | 56224 |
Gene Name: | tetraspanin 5 |
Gene Aliases: |
2810455A09Rik, 4930505M03Rik, NET-4, Tm4sf9 |
Location: |
Chr.3:138742208-138904433 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan5 | NM_019571.5 | NP_062517.1 | ||
| NR_121195.1 | ||||
| AF121344.1 | AAF28869.1 | |||
| AK013350.1 | BAB28804.1 | |||
| AK015705.1 | ||||
| AK033543.1 | ||||
| AK187703.1 | ||||
| BC046784.1 | ||||
| BC058695.1 | AAH58695.1 |

Genomic Map