Assay Details
Target Gene Details
Entrez Gene ID: | 76947 |
Gene Name: | NADH dehydrogenase (ubiquinone) complex I, assembly factor 6 |
Gene Aliases: |
2310030N02Rik |
Location: |
Chr.4:11051042-11076235 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf6 | NM_001085493.2 | NP_001078962.1 | ||
| AK009547.1 |

Genomic Map