Assay Details
Target Gene Details
Entrez Gene ID: | 66264 |
Gene Name: | coiled coil domain containing 28B |
Gene Aliases: |
1110002E08Rik, 1810010N17Rik |
Location: |
Chr.4:129619274-129623908 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc28b | NM_025455.2 | NP_079731.2 | ||
| AK003287.1 | BAB22692.1 | |||
| AK007427.1 | BAB25032.1 | |||
| AK009176.1 | BAC25244.1 | |||
| AK028262.1 | BAC25848.1 | |||
| AK031278.1 | 1 | 143 | ||
| AK158016.1 | BAE34319.1 | |||
| BB848581.1 | ||||
| BC028873.1 | AAH28873.1 | |||
| BQ031123.1 |
More Information
Set Membership: |
|

Genomic Map