Assay Details
Target Gene Details
Entrez Gene ID: | 209584 |
Gene Name: | tRNA-yW synthesizing protein 3 homolog (S. cerevisiae) |
Gene Aliases: |
5230400J09Rik |
Location: |
Chr.3:154576520-154597098 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyw3 | NM_001168358.1 | NP_001161830.1 | ||
| NM_172474.5 | 5 | 564 | NP_766062.1 | |
| AK030344.1 | 5 | 562 | BAC26913.1 | |
| AK034820.1 | 5 | 549 | BAC28842.1 | |
| BC086686.1 | 5 | 538 | ||
| BC087879.1 | 5 | 528 | AAH87879.1 | |
| BC141040.1 | 5 | 543 |
More Information
Set Membership: |
|

Genomic Map