Assay Details
Target Gene Details
Entrez Gene ID: | 51813 |
Gene Name: | cyclin C |
Gene Aliases: |
AI451004, AU020987, CG1C |
Location: |
Chr.4:21727701-21759922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnc | NM_001122982.2 | NP_001116454.1 | ||
| NM_001290420.1 | NP_001277349.1 | |||
| NM_001290422.1 | NP_001277351.1 | |||
| NM_016746.4 | NP_058026.2 | |||
| AK009615.1 | ||||
| AK014079.1 | ||||
| AK037957.1 | BAC29908.1 | |||
| AK135472.1 | BAE22544.1 | |||
| AK153035.1 | BAE31667.1 | |||
| BC003344.1 | ||||
| BC029211.1 | ||||
| BC037689.1 | ||||
| BC062376.1 | AAH62376.1 | |||
| BC087544.1 | AAH87544.1 | |||
| BC100396.1 | AAI00397.1 | |||
| BC120649.1 | AAI20650.1 | |||
| BC120677.1 | AAI20678.1 | |||
| U62638.1 | AAB05260.1 |
More Information
Set Membership: |
|

Genomic Map