Assay Details
Target Gene Details
Entrez Gene ID: | 114143 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit B |
Gene Aliases: |
2310024H13Rik, Atp6f, VMA16 |
Location: |
Chr.4:117884326-117887340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0b | NM_033617.3 | NP_291095.1 | ||
| AB060654.1 | BAB61954.1 | |||
| AF356006.1 | AAL02096.1 | |||
| AK009495.1 | ||||
| AK021272.1 | ||||
| AK151365.1 | ||||
| AK151847.1 | BAE30738.1 | |||
| AK152330.1 | BAE31129.1 | |||
| AK152823.1 | BAE31524.1 | |||
| AK170493.1 | 1 | 1486 | ||
| BC009169.1 | AAH09169.1 | |||
| BC030393.1 | AAH30393.1 |
More Information
Set Membership: |
|

Genomic Map