Assay Details
Target Gene Details
Entrez Gene ID: | 114143 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit B |
Gene Aliases: |
2310024H13Rik, Atp6f, VMA16 |
Location: |
Chr.4:117884326-117887340 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0b | NM_033617.3 | 7 | 593 | NP_291095.1 |
| AB060654.1 | 7 | 554 | BAB61954.1 | |
| AF356006.1 | 7 | 597 | AAL02096.1 | |
| AK009495.1 | 7 | 584 | ||
| AK021272.1 | 7 | 573 | ||
| AK151365.1 | 7 | 564 | ||
| AK151847.1 | 7 | 581 | BAE30738.1 | |
| AK152330.1 | 7 | 598 | BAE31129.1 | |
| AK152823.1 | 7 | 581 | BAE31524.1 | |
| AK170493.1 | 1 | 2125 | ||
| AK189647.1 | ||||
| AK205813.1 | ||||
| AK206535.1 | ||||
| AK208636.1 | ||||
| AK210235.1 | ||||
| AK211031.1 | ||||
| AK211051.1 | ||||
| AK212370.1 | ||||
| BC009169.1 | 7 | 594 | AAH09169.1 | |
| BC030393.1 | 7 | 578 | AAH30393.1 |
More Information
Set Membership: |
|

Genomic Map