Assay Details
Target Gene Details
Entrez Gene ID: | 12448 |
Gene Name: | cyclin E2 |
Gene Aliases: |
- |
Location: |
Chr.4:11189214-11204779 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccne2 | NM_001037134.2 | 6 | 598 | NP_001032211.1 |
| NM_001282943.1 | 6 | 511 | NP_001269872.1 | |
| NM_009830.3 | 6 | 508 | NP_033960.1 | |
| AF091432.1 | 5 | 403 | AAC80527.1 | |
| AF106691.1 | 4 | 378 | AAD08817.1 | |
| AK010929.1 | 6 | 438 | ||
| AK011883.1 | 1 | 70 | BAB27897.1 | |
| AK031116.1 | 6 | 435 | ||
| AK037791.1 | 6 | 438 | BAC29873.1 | |
| AK158720.1 | 6 | 1024 | BAE34625.1 | |
| AK167428.1 | 6 | 438 | BAE39516.1 | |
| BC053727.1 | 5 | 398 | AAH53727.1 |
More Information
Set Membership: |
|

Genomic Map