Assay Details
Target Gene Details
Entrez Gene ID: | 12448 |
Gene Name: | cyclin E2 |
Gene Aliases: |
- |
Location: |
Chr.4:11189214-11204779 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccne2 | NM_001037134.2 | NP_001032211.1 | ||
| NM_001282943.1 | NP_001269872.1 | |||
| NM_009830.3 | NP_033960.1 | |||
| AF091432.1 | AAC80527.1 | |||
| AF106691.1 | AAD08817.1 | |||
| AK010929.1 | ||||
| AK011883.1 | BAB27897.1 | |||
| AK031116.1 | ||||
| AK037791.1 | BAC29873.1 | |||
| AK158720.1 | BAE34625.1 | |||
| AK167428.1 | BAE39516.1 | |||
| BC053727.1 | AAH53727.1 |
More Information
Set Membership: |
|

Genomic Map