Assay Details
Target Gene Details
Entrez Gene ID: | 230098 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 39 |
Gene Aliases: |
E130306D19Rik |
Location: |
Chr.4:43496139-43508769 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef39 | NM_001013377.2 | 2 | 239 | NP_001013395.2 |
| AK087493.1 | 2 | 192 | ||
| AK153880.1 | 2 | 234 | ||
| BC080700.1 | 2 | 211 | AAH80700.1 | |
| BY009353.1 | 2 | 241 |
More Information
Set Membership: |
|

Genomic Map