Assay Details
Target Gene Details
Entrez Gene ID: | 14590 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
gamma-GH |
Location: |
Chr.4:20041987-20066111 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggh | NM_010281.2 | NP_034411.2 | ||
| AF051102.1 | AAC70002.1 | |||
| AK089996.1 | ||||
| BB610405.1 | ||||
| BC009809.1 | AAH09809.1 | |||
| BC150898.1 | ||||
| BE859353.1 |
More Information
Set Membership: |
|

Genomic Map