Assay Details
Target Gene Details
Entrez Gene ID: | 230233 |
Gene Name: | inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein |
Gene Aliases: |
3110040G09Rik, 6030413P05, C78473, Elp1, IKAP |
Location: |
Chr.4:56749680-56802331 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkap | NM_026079.3 | 13 | 1588 | NP_080355.2 |
| AF140786.1 | 13 | 1516 | AAO15309.1 | |
| AF367244.1 | 13 | 1544 | AAL36025.1 | |
| AF387811.1 | 13 | 1555 | AAL40925.1 | |
| AK031367.1 | 13 | 1533 | ||
| AK035944.1 | 13 | 1590 | BAC29252.1 | |
| AK147153.1 | 13 | 1591 | BAE27720.1 | |
| AK165168.1 | 10 | 1065 | ||
| BC052387.1 | 13 | 1500 | AAH52387.2 |
More Information
Set Membership: |
|

Genomic Map